Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Fischertechnik Pneumatic Processing Center
77577 520529
470210-856EA 989.99 USD InStock
470210-856 470117-586
Fischertechnik Pneumatic Processing Center
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Machining center with bin for workpieces, rotary table, processing station, conveyor belt for transporting work pieces and model compressor. Contains two double-acting and one single-acting pneumatic cylinder, 5 x 3/2 solenoid valves, two DC motors, four proximity sensors, and four light barriers consisting of phototransistor and lens tip bulb. Model is mounted on a stable wooden board. Includes eight digital inputs and seven 9V outputs. Unit dimensions are 450 x 410 x 190 mm.
Ordering information: Pneumatic processing centers require Fischertechnik Energy Set, available separately.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty