Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Baritainer® Jerry Can, Dynalon
Supplier: Dynalon
Baritainer®
405594-0004 405594-0001 405594-2070 405594-0002
89508-632EA 35.79 USD InStock
89508-632 89508-634 89508-638 89508-636
Baritainer® Jerry Can, Dynalon
Bottles Jerricans
Baritainer® Jerry Cans are constructed of HDPE and Quoral®, a barrier resin which provides an oxygen and solvent barrier that is up to 5 times better than fluorination
Store a large volume of liquid in these compact, durable, impact- and dent-resistant, rustproof, economical Baritainer® Jerry Cans. An integral handle and lightweight construction make them easy to carry while the notched bottom makes them easy to stack.
Baritainer® Jerry Cans are available in natural HDPE and feature a black or natural tamper-evident, lined, leak proof HDPE closure system. FDA compliant. Recyclable.
They are a perfect substitute for more expensive steel or stainless steel containers when a barrier for flavors, ingredients, odors, and organic solvents is needed.
Certifications: The 4L, 5L, and 10L containers are rated UN 3H1/X1.2/250. The 20L container is rated UN 3H1/Y1.8/100.
Ordering information: Accessories sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty