Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

alamarBlue® reagent Cell Proliferation Assay

Supplier: BIO-RAD LABORATORIES INC MS

BUF012B BUF012A
100234-636EA 442 USD InStock
100234-636 100234-634
alamarBlue® reagent Cell Proliferation Assay
alamarBlue® reagent
alamarBlue® is a cell proliferation assay that provides a rapid, sensitive and economical way to quantitatively measure cell proliferation and cytotoxicity in various human and animal cell lines. It can also be used to study mycobacteria, bacteria and fungi.
The following features and benefits establish alamarBlue as a preferred alternative reagent to similar cell proliferation assays (MTT, XTT, 3H Thymidine or neutral red) which will help you to get the results you need:

  • Simple : Water soluble, just add and measure the formula in cell cultures
  • Flexible: Colorimetric and fluorometric detection, for both suspended and attached cell lines
  • Non-toxic: alamarBlue does not kill cells and is safe to the user and the environment
  • Reliable: Highly referenced for cytotoxicity and viability assays
  • Scalable: Effortless scale up for high throughput assays
  • High Sensitivity: Detects as few as 50 cells
  • Stable: Proprietary buffering agents ensures alamarBlue is suitable for time course studies
  • Economical: No cell lysis allows cells to be cultured for subsequent assays

alamarBlue® can be used for applications such as functional assay, immunofluorescence and ELISA. This reagent is offered in two sizes:
25 mL = Enough for 2500 wells /96-well plate
100 mL = Enough for 10000 wells /96-well plate
Note: Calculations assume 100 µl final volume per well (96-well plate).


Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty