Foam Microtube Rack
Supplier: Heathrow Scientific
HS4283P
89194-766CS 958.51 USD
89194-766
Foam Microtube Rack
Racks Tube Racks
Lightweight, small footprint microtube rack is ideal for long-term studies or tube storage.
  • Two-piece racks are constructed of expanded polystyrene (EPS) foam
  • Extra spacing between wells allows easy gripping with fi ngertips
  • Racks stack securely with nesting features on lid and base
Store down to temperatures of –80 °C.