Silica gel 6-20 mesh, with indicator

ANAC79085-580 ANAC79085-760 ANAC79085-380
CA71008-406EA 1160.44 USD
CA71008-406 CA71008-408 CA71008-404
Silica gel 6-20 mesh, with indicator
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR