Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

2-Ethoxyethanol ≥99.5%

Danger
Synonyms: CellosolveEthylene glycol monoethyl ether

065V-07 195V-06
CA71007-196EA 1138.46 USD InStock
CA71007-196 CA71007-192
2-Ethoxyethanol ≥99.5%
2-Ethoxyethanol


Formula: C₂H₅OCH₂CH₂OH
MW: 90.12 g/mol
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.932 g/cm³ (20 °C)
Flash Pt: 40 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002869
CAS Number: 110-80-5
EINECS: 203-804-1
UN: 1171
ADR: 3,III
REACH: 01-2119560582-38
Merck Index: 13,03786
https://us.vwr.com/stibo/web/std.lang.all/93/02/2299302.jpg
Order Now
Specification Test Results
Assay 99.5% min.
Water (H2O) 0.1% max.

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty