Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eriochrome Black T, Powder indicator for complexometry

Warning
Synonyms: Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonateEBTSolochrome Black TSodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulfonate

ANAC37835-080
CA11021-970EA 182.76 USD InStock
CA11021-970
Eriochrome Black T, Powder indicator for complexometry
Eriochrome Black T
Colour Index: 14645


Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
EINECS: 217-250-3
UN: 3077
ADR: 9,III
Merck Index: 12,03709
Order Now
Specification Test Results
C.I. 14645 Mordant black 11
Appearance: powder Passes Test
Clarity of solution Passes Test
Suitable as complexometric indicator Passes Test

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty