Agar, powder, Lab-Grade

Synonyms: Agar Agar

ANAC02116-380
CA71006-520EA 687.14 USD
CA71006-520
Agar, powder, Lab-Grade
Agar
Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://us.vwr.com/stibo/web/std.lang.all/81/91/2298191.jpg

Order Now

Specification Test Results

Loss on drying at 105°C 20% max.
Total ash 6.5% max.
pH info only

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR