Would you like to log in again?
Professional LYSOL® No Rinse Sanitizer
Supplier: T FRANK MCCALLS INC
Lysol®
36241-74389
21899-626CS 166.28 USD InStock
21899-626
Professional LYSOL® No Rinse Sanitizer
Cleaning Agents Disinfectants
Meets federal food code requirements for a no rinse sanitizer
Formulated for three-bin sink and clean-in-place applications. Also used to disinfect and deodorize. Controls algae in swimming pools and industrial non-potable water supplies. Economical, concentrated formula makes up to 512 gallons.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty