Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
VWR® Traceable® Instant-Recall Memory Timer
5122
89370-114EA 24.93 USD InStock
89370-114
VWR® Traceable® Instant-Recall Memory Timer
Timers Digital Timers
Input a time and the instant memory always returns the display to the programmed time.
  • Memory function favors use with one piece of equipment
  • Ideal for routinely timed tests
  • Instant memory programming
Designed specifically for a routine test, quality control requirements, and repetitive analysis, it is the lab workhorse
This is the ideal timer to dedicate to a routinely timed test. Bright, extra-large, 5/8" high digits count down at any set time from 99 minutes 59 seconds to zero. At zero a loud 30-second alarm sounds.
The unique memory feature encourages devoting this timer to one piece of laboratory equipment such as a water bath, oven, stirrer, or incubator.
Certifications: To ensure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibrated laboratory accredited by A2LA. It indicates traceability to NIST.
Ordering information: Supplied with a magnetic back, flip-open stand, and Traceable® Certificate.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty