Would you like to log in again?
VWR® Traceable® Wireless Radio-Signal Refrigerator Thermometer
Supplier: VWR International
VC6424 VC1004 VC2004 VC2060 VC1060 VC6425 6424
10171-322EA 110.99 USD InStock
10171-322 10171-380 10171-520 10171-602 10171-486 10171-216 89369-160
VWR® Traceable® Wireless Radio-Signal Refrigerator Thermometer
Thermometers Digital Thermometers Digital Thermometers for Special Applications
Unit wirelessly reads refrigerator temperatures up to 100 feet away.
  • Insulated from rapid temperature changes
  • Signal penetrates refrigerators and walls
  • Memory feature stores minimum/maximum readings
  • User programmable alarm
  • Connected to up to three remote sensors
Temperature readings from up to three remote refrigerator sensors are sent to a main unit via radio signal (frequency of 433 MHz) penetrates refrigerators and walls. Sealed bottle is insulated from rapid temperature changes when refrigerator door is opened. Liquid in sealed bottle is GRAS - generally recognized as safe by the US FDA. Data is updated every 30 seconds. Minimum/Maximum memory allows monitoring conditions over any time period. Distinctive alarm sounds when temperature rises above or falls below settable high and low set points. Alarm may be conveniently set in 1° increments. Indicator on main unit shows rising/falling temperature changes.
Certifications: To ensure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibrated laboratory accredited by A2LA. It indicates traceability to NIST.
Ordering information: Supplied with one main unit, one remote unit, flip-open stand, wall mount, Velcro®, batteries and Traceable® Certificate.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to VWR's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty