2X YT Broth Powder, Grow Cells
MBPE-5016 MBPE-5032
101385-044EA 56.29 USD InStock
101385-044 101385-046
2X YT Broth Powder, Grow Cells
Media for Microbiology Dehydrated Media for Microbiology
2X YT Broth is a highly enriched medium developed to improve yield in plasmid bearing Escherichia coli.
To Prepare: 1 X solution, add 31 g/1000 mL DI water and swirl to create a suspension. Steam sterilize at 15 psi for 20 minutes at 121º C.