Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

Supplier: GROWCELLS MS
Synonyms: BCIG

MESP-5003
101385-652EA 545.37 USD InStock
101385-652
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Chromogenic substrate for B-glactosidase

Cells with active B-galactosidase will produce a blue precipitate upon hydrolysis. It is the substrate of choice for blue-white selection of recombinant bacterial colonies. The X-gal quality and purity is essential to obtain high-quality reliable results, therefore our ultra pure molecular grade X-gal is optimized for use from the best raw materials


Formula: C₁₄H₁₅BrClNO₆
MW: 408,63 g/mol
Melting Pt: 237 to 239 °C
Storage Temperature: -20°C
MDL Number: MFCD00005666
CAS Number: 7240-90-6
EINECS: 230-640-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty