Would you like to log in again?
LB Lennox Broth Powder, Grow Cells
MBPE-2020 MBPE-2050 MBPE-2100
101385-014EA 55.14 USD InStock
101385-014 101385-016 101385-018
LB Lennox Broth Powder, Grow Cells
Media for Microbiology Dehydrated Media for Microbiology
Lysogeny broth (LB) - Lennox formula is a nutrionally rich medium optimized for culture of transformed E.coli
To Prepare: 1 X solution, add 20 g/1000 ml DI water and swirl to create a suspension. Steam sterilize at 15 psi for 20 minutes at 121º C.
The lower salt in Lennox's formulation is ideal for salt-sensitive antibiotics.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty