LB Miller's Broth Powder, Grow Cells
MBPE-1050 MBPE-1100 MBPE-1020
101384-998EA 54.85 USD
101384-998 101385-002 101385-004
LB Miller's Broth Powder, Grow Cells
Lysogeny broth (LB) - Miller's formula is a nutrionally rich medium optimized for culture of transformed E.coli.
To Prepare: 1 X solution, add 25 g/1000 ml DI water and swirl to create a suspension. Steam sterilize at 15 psi for 20 minutes at 121º C.