Multipoint Fuel Injection Simulator
AT5101
470210-724EA 2298.99 USD
470210-724
Multipoint Fuel Injection Simulator
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This training simulator is a comprehensive instructional module that plugs directly into the Advanced Autotronics Simulator workstation, designed to train students on modern multipoint fuel injection systems.
The topics covered on this board include: MAP and MAF sensors, motorized throttle, injectors, DIS ignition coils, CMP: Hall Effect camshaft position sensor, CKP: inductive crankshaft position sensor, knock sensor, accelerator position Hall Effect sensor, engine electronic control unit (ECU), and troubleshooting exercises. The simulator comes with interactive courseware which contains essential theory enhanced with vivid simulations, guided exercises that interact with the Multipoint Fuel Injection simulator, guided diagnostic exercises and self assessment exercises.