Fluid A, Hardy Diagnostics
Supplier: Hardy Diagnostics
U342 U342
89498-954PK 641.76 USD
89498-954 CA89498-954
Fluid A, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Fluid A, USP, 300ml fill, 500ml glass bottle with needle-port septum screw cap, order by the package of 10, by Hardy Diagnostics
For use as a diluting and rinsing fluid per USP procedures.
Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.
Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center
Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.
Certifications: USP