SP4 Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
U191
CA89498-950
SP4 Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
SP4 Broth with Glucose, 100ml fill, 4 ounce boston round bottle, order by the each, by Hardy Diagnostics. MINIMUM ORDER: 10 each.
For the cultivation and differentiation of mycoplasmas.
SP4 Broth with Glucose, 100ml fill, 4 ounce boston round bottle, order by the each, by Hardy Diagnostics
MINIMUM ORDER: 10 each.