D/E Neutralizing Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
U113 U113
89423-718PK 317.73 USD
89423-718 CA89423-718
D/E Neutralizing Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
D/E Neutralizing Agar, 300ml, 16 ounce boston round bottle, order by the package of 12, by Hardy Diagnostics.
For the neutralization and testing of antiseptics and disinfectants.