Violet Red Bile Agar (VRBA) with MUG, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
J132 J132
89497-602PK 49.14 USD InStock
89497-602 CA89497-602
Violet Red Bile Agar (VRBA) with MUG, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Violet Red Bile Agar (VRBA) with MUG, 10ml fill, 15x100mm biplate, order by the package of 10, by Hardy Diagnostics.
For the detection of coliforms in food or dairy products. MINIMUM ORDER: 10 packs.
Ordering information: MINIMUM ORDER: 10 packs.