    Sort by:
  • Search Within Results
2-(Methylthio)benzonitrile
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   2-Cyanothioanisole 2-(Methylmercapto)benzonitrile. Grade:98, Melting Point C37-39*. Boiling Point C:116-117*/5mm. C8H7NS. 6609-54-7. HARMFUL