You Searched For: (R)-(-)-2-Aminohexane
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Thermo Scientific Chemicals
Description: (2R)-Hexylamine (R)-1-Methylpentylamine. Grade: ChiPros\(tm 99+%, ee 96+. Melting Point C<-20*. Boiling Point C: 116*. C6H15N. 70492-67-0. Trademark of BASF AG. FLAMMABLE CORROSIVE HARMFUL AIR SENSITIVE