    Sort by:
  • Search Within Results
You Searched for: "trimethylsilyl-isothiocyanate"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Grade: 94, Melting Point C-33*. Boiling Point C: 143-144*. C4H9NSSi. 2290-65-5. FLAMMABLE TOXIC LACHRYMATORY KEEP COLD MOISTURE SENSITIVE