Would you like to log in again?
Timing Car
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SV681 SV683
Replace your air track and photogate timers with this unique device that allows students to investigate kinematics and dynamic motion without the need for a photogate.
Perform several Laws Of Motion experiments with the timing car that features built-in motion and direction detection with a two-channel sensor, bright LCD display, remote detection sensor, a user replaceable 9V battery and an intelligent micro-controller based circuitry to sense millimeter level movement of the car on linear and non-linear motion. Combine with additional accessories and ramps for added investigation, available separately.
ORDER
Expand All / Collapse All
Timing Car
Description Supplier No. VWR Catalog Number Unit Price Quantity
Timing Car SV681 470211-158 Case of 10 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Inclined Accessories for Plan Ramp SV701 470211-166 Case of 10 Retrieving
Each Retrieving
Plane Ramp for Timing Car SV682 470211-162 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty