Programmable DC Electronic Load, 600W
8512
470210-678EA 3056.99 USD
470210-678
Programmable DC Electronic Load, 600W
The affordable, laboratory grade load is well suited for testing and evaluating a wide variety of DC power sources.
  • Extensive Protection
  • Flexible operating modes
  • excellent measurement accuracy
This DC electronic load can operate in CC, CV, CR or CP mode while voltage/current or resistance/power values are measured and displayed in real time and is fully programmable via RS232 or USB interface. Extensive protection, including over temperature, over power, over voltage, over current and reverse polarity will help protect your valuable prototype. This DC electronic load is a great choice for characterizing DC Power supplies, DC-DC Converters, batteries, fuel cells and solar cells.