Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
VWR® LX Immersion Circulator
LXC-9V11B
89501-480EA 670.92 USD InStock
89501-480
VWR® LX Immersion Circulator
Circulating Baths Open Circulator Systems Thermostat Controllers
Create your own circulating bath system using your own vessel or a VWR® open bath tank.
  • Space-saving design
  • Backlit LCD display
  • Built-in timer
Large backlit LCD display allows you to monitor set-point and actual temperature from across the lab. The adjustable mounting clamp fastens securely to both flat and rounded tank walls, while the integral pump/heating coil protection shields pump and heater, yet offers easy access to cleaning. The streamlined design maximizes available reservoir space, while the built-in timer alerts you when samples are ready or processes are complete.
Certifications: 120V unit is ETL listed (North America only).
Ordering information: 240V/50Hz model is also available; contact your VWR sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty