Would you like to log in again?
BD BBL™ Enteropluri Test, BD Diagnostics
Supplier: BD
215336 215336
BD215336PK 478.74 USD InStock
CA11027-062 BD215336
BD BBL™ Enteropluri Test, BD Diagnostics
Microbiology Test Systems Antigen-Antibody Tests
The BBL Enteropluri Test is a 12-sector system containing special culture media that permits identification of the Enterobacteriaceae and other gram negative, oxidase negative bacteria
The system allows the simultaneous inoculation of all media present in the sectors and the execution of 15 biochemical reactions. Microorganism is identified evaluating the color change of the different culture media after 18-24 hours of incubation at 36 ± 1°C and by a code number obtained from biochemical reaction interpretation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty