Would you like to log in again?
Model SI-7 Silica Test Kit, Hach
Supplier: Hach
2255000 2255000
89500-714EA 162.64 USD InStock
89500-714 CA89500-714
Model SI-7 Silica Test Kit, Hach
Environmental Test Kits
Dissolved silica is found in almost all natural waters
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Its presence is undesirable for many industrial uses because of scale formation. High-pressure boiler turbines are especially susceptible. This simple, low-cost color disc method is the most accurate visual test kit method. This kit is popular in many industrial applications, such as boiler turbines that are susceptible to scale. Provides fast results with an easy-to-carry test kit.
Ordering information: Kit includes three reagents, viewing tube, color comparator box, color discs, viewing adapter, bottle, stopper, instruction sheet, and carrying case.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty