Would you like to log in again?
Model PO-23 Phosphate Test Kit, Hach
Supplier: Hach
224902 224902
89500-710EA 193.19 USD InStock
CA89500-710 89500-710
Model PO-23 Phosphate Test Kit, Hach
Environmental Test Kits
Simple, low-cost color disc method is the most accurate visual test kit method
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Achieve fast results with this easy-to-carry test kit.
Ordering information: Kit includes three reagents, color disc, comparator box, two viewing tubes, bottle, dropper tube, stove and cover, test tube clamp, deionized water, instructions and carrying case.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty