Would you like to log in again?
Blue X-Ray Film, Phenix Research Products
Supplier: THOMAS SCIENTIFIC HOLDINGS, LLC MS
F-BX810 F-BX57
101100-268PK 121.38 USD InStock
101100-268 101100-098
Blue X-Ray Film, Phenix Research Products
Autoradiography Films Films
Blue X-Ray Film provides sharp contrast and resolution for sequencing procedures, and is ideal for diverse applications using P32 and I125, single or double emulsion.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty