Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Silica Test Kit, Model SI-5, Hach
Supplier: Hach
1455400 1455400
89500-644EA 180.16 USD InStock
CA11024-958 89500-644
Silica Test Kit, Model SI-5, Hach
Environmental Test Kits
Dissolved silica is found in almost all natural waters
  • Popular in many industrial applications, such as boiler turbines that are susceptible to scale
  • Provides fast results with an easy-to-carry test kit
  • Simple, low-cost color disc method is the most accurate visual test kit method
Hazardous Items; Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Its presence is undesirable for many industrial uses because of scale formation. High-pressure boiler turbines are especially susceptible. This simple, low-cost color disc method is the most accurate visual test kit method.
Ordering information: Supplied with four reagents, deionized water, viewing tube, stopper, color comparator box, color discs, bottle dropper, instruction sheet, and carrying case.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty