Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Detergents Test Kit, Model DE-2, Hach
Supplier: Hach
143203 143203
89500-636EA 374.69 USD InStock
CA89500-636 89500-636
Detergents Test Kit, Model DE-2, Hach
Environmental Test Kits
The presence of detergents in surface or ground waters indicates industrial or domestic pollution
  • Simple, low-cost color disc method is the most accurate visual test kit method
  • Achieve fast results with an easy-to-carry test kit
Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
In natural waters anionic LAS and ABS detergents cause excessive foaming, produce undesirable taste and odors, promote growth of disease-producing organisms, and tend to kill aquatic life. This kit's simple, low-cost color disc method is the most accurate visual test kit method.
Ordering information: Includes four reagents, one thimble, two test tubes, one rubber bulb, two glass tubes, glass wool, one dropper, two stoppers, a color comparator box, one color disc, instructions, and a carrying case.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty