Bovine Fibronectin from Plasma
Supplier: MP Biomedicals
0215002501
IC15002501EA 304 USD
IC15002501
Bovine Fibronectin from Plasma
Proteins and Peptides
Soluble in water: soluble1 mg/mL at 37 °C.
Fibronectin are adhesive glycoproteins and are found both as cell surface proteins and in plasma. They are large glycoproteins found in cells and extracellular fluids. They are involved in a wide range of biological activities including cell adhesion to extracellular matrices, blood coagulation and wound healing. In tissue culture, fibronectins promote cell adhesion and spreading on synthetic surfaces. Certain cell types show morphological changes relating to loss of fibronectin from the cell surface or depletion of fibronectin from the culture medium. Fibronectins are of two, closely related, major types: cellular fibronectin, otherwise known as Large External Transformation Sensitive (LETS) protein and plasma fibronectin, also called Cold-insoluble Globulin (CLg).
Fibronectin has been shown to stimulate the proliferation of rat osteoblasts in vitro; to induce odontoblast polarization and to regulate the differentiation of human dental pulp cells into odontoblasts. It has wide applications in cellular research into proliferation, differentiation, development and morphology. Precoating of glass, plastic and micro-carrier beads aids cell attachment, particularly those cell which are deficient in cell surface fibronectin. It has been reported that proteolytic digests of fibronectin are mitogenic for CH23 fibroblasts.
Fibronectin is commonly used under reduced-serum and serum-free culture conditions.