Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Varispenser® and Varispenser® plus Bottle Top Reagent Dispenser, Eppendorf

Supplier: Eppendorf
4961000063 4960000027 4961000020 4960000019 4960000035 4961000039 4960000060 4961000055 4961000012 4960000051 4961000047
Varispenser® adjustable-volume bottle-top reagent dispenser features a patented safety valve for de-airing without loss of reagent and a channel-sealing cap that prevents media contact; no crystallization of liquid, no dripping when valve closed

Allows fine adjustment depending on the viscosity of the liquid, with a rapid volume setting using a precise graduation scale. It is, autoclavable and extremely chemical-resistant.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Volume Supplier No. VWR Catalog Number Unit Price Quantity
Varispenser® Size 2 1-5 mL 4960000027 89133-950 Each Retrieving
Product available on GSA Advantage®
Varispenser® Size 6 20-100 mL 4960000060 89133-958 Each Retrieving
Product available on GSA Advantage®
Varispenser® plus Size 1 0.5-2.5 mL 4961000012 89133-960 Each Retrieving
Product available on GSA Advantage®
Varispenser® plus Size 2 1-5 mL 4961000020 89133-962 Each Retrieving
Product available on GSA Advantage®
Varispenser® plus Size 3 2-10 mL 4961000039 89133-964 Each Retrieving
Product available on GSA Advantage®
Varispenser® plus Size 4 5-25 mL 4961000047 89133-966 Each Retrieving
Product available on GSA Advantage®
Varispenser® plus Size 5 10-50 mL 4961000055 89133-968 Each Retrieving
Product available on GSA Advantage®
Varispenser® plus Size 6 20-100 mL 4961000063 89133-970 Each Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
PTFE Thread Adapter, 32 - 28 mm 4960835005 89399-914 Each Retrieving
PTFE Thread Adapter, 32 - 38 mm 4960836001 89399-912 Each Retrieving
PTFE Thread Adapter, 32 - 45 mm 4960837008 89399-918 Each Retrieving
PTFE Thread Adapter, 38 - 32 mm 4960838004 89399-920 Each Retrieving
PTFE Thread Adapter, 45 - 38 mm 4960839000 89399-916 Each Retrieving
PTFE Thread Adapter, 45 - 40 mm 4960834009 89399-922 Each Retrieving
Spiral Discharge Tube, 80 cm, for Varispenser® and Varispenser® plus 4960825000 75874-630 Each Retrieving
Spiral Discharge Tube, 80 cm, for Varispenser® and Varispenser® plus, 2.5 mL, 5 mL and 10 mL 4960824003 75874-628 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty