Would you like to log in again?
Single Position Magnetic Stirrer, BeLLCo Glass
Supplier: BELLCO GLASS INC MS
7760-06000
Single position magnetic stirrer with a 30–1000rpm speed range
ORDER
Expand All / Collapse All
Description Electrical Overall Dimensions Maximum Capacity Speed Range Supplier No. VWR Catalog Number Unit Price Quantity
Single Position Magnetic Stirrer 115V, 60Hz, 1.0A 16W x 16D x 13H cm (6.3 x 6.3 x 5.1") 1 L (33.8 oz.) 30–1000 rpm 7760-06000 100487-316 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty