Would you like to log in again?
AxyMat™ Silicone Sealing Mats, Axygen Scientific
Supplier: Corning
Axygen® AxyMat™
AM-2ML-RD-S AM-2ML-RD AM-2ML-SQ-IMP AM-750UL-RD AM-750UL-RD AM-48 AM-24-SQ AM-500UL-RD-S AM-500UL-SL AM-2ML-SQ-S AM-2ML-RD-IMP AM-500UL-SL AM-2ML-RD AM-500UL-RD AM-2ML-RDIMP
10011-946CS 916.86 USD InStock
10011-946 CA10011-126 80078-816 47734-658 CA47734-658 89179-614 89179-610 80078-828 80089-654 89179-612 80078-404 CA80089-654 10011-126 47743-826 CA80078-404
AxyMat™ Silicone Sealing Mats, Axygen Scientific
Seals Microplate Sealing Mats
Sealing mats minimize evaporation in a wide range of temperatures in PCR and storage applications
AxyMats™ are chemically resistant, reusable sealing mats, that provide a high degree of confidence. For storage applications involving more aggressive solvents, they contain no adhesives or extractable to contaminate samples. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty