Corning® High Concentration Synthemax II Microcarriers
3784
89499-406CS 243.11 USD
89499-406
Corning® High Concentration Synthemax II Microcarriers
Corning Synthemax II Microcarriers are designed to simplify scale-up for stem cells
The Low Concentration Synthemax II microcarriers is optimized for hMSC culture. The High Concentration Synthemax II microcarriers is optimized for hESC or hiPSC culture.
All Corning Microcarriers are polystyrene based.
The microcarriers are sterile and ready-to-use for culture. The Synthemax II surface is Corning's unique animal-free, synthetic peptide surface that mimics the natural cell environment. This surface supports the adhesion and expansion of stem cells in their undifferentiated state and directed differentiation into specialized cell types.