Stablcal® Turbidity Standard, 300 mNTU, 1000 mL, Hach
Supplier: Hach
Stablcal®
2697953 2697953
97026-470EA 186.39 USD InStock
97026-470 CA11016-442
Stablcal® Turbidity Standard, 300 mNTU, 1000 mL, Hach
Standards Turbidity Standards
  • Precise, Pre-mixed Formazin Solutions
  • Stablcal is Safe to Handle
  • No special glassware or difficult lab techniques
  • Developed for use in any turbidimeter.
  • Enhanced Stability Saves Time and Labor
Stablcal® Formazin Turbidity Standard, Bulk Suspensions. 0.30 NTU. Bottle/1000 mL.