Radiometer Analytical PHC3081-6 Combination pH Electrode with Temperature Sensor, Glass Body, Hach
E16M344
89174-270EA 560.4 USD
89174-270
Radiometer Analytical PHC3081-6 Combination pH Electrode with Temperature Sensor, Glass Body, Hach
The Radiometer PHC3081-6 Combination pH Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat
  • The right probe for your application.
  • Glass sensors made for longlife and durable use.
The PHC3081-6 has a fixed cable with BNC + RJ9 connectors for use with Hach sensION+ benchtop pH meters.
AgCl), integrated temperature sensor, and porous pin junction for general purpose applications.
Ordering information: Radiometer Analytical PHC3081-6 combination pH electrode with Certificate of Conformity and small bottle of internal electrolyte solution included.