Would you like to log in again?
AlphaTec® 58-535, Ansell Grip® Technology Nitrile Gloves, Knit Lined, Ansell
Supplier: Ansell Healthcare
219008 219009 219007
89011-899CS 953.72 USD InStock
89011-899 89011-901 89011-897
AlphaTec® 58-535, Ansell Grip® Technology Nitrile Gloves, Knit Lined, Ansell
Gloves
This glove has revolutionary Ansell Grip Technology – a nitrile layer filled with microscopic channels that actively transfer oils and other lubricants away from the gripping surface.
  • Oil channeled away by microscopic channels
  • Superior tactility and dexterity
  • Consistent polymer coating preventing chemical leakage
Its consistent polymer coating significantly reduces the likelihood of chemical leakage
Ideal uses are in chemical plants, transportation, chemical processing, oil refining, petrochemicals, cleaning and de-greasing, handling equipment and abrasive materials coated with oil, grease, acids, solvents, handling fillers, dye agents, cleaning solvents, cement and mortar mixing.
It has a unique combination of grip, chemical resistance and superior comfort, featuring a gauntlet-style cuff and knit lining. It has superior tactility and dexterity for greater control, significantly reducing fatigue and Carpel Tunnel Syndrome-related injuries.
Ordering information: Also available in a shorter 12" length, see AlphaTec 58-530.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty