Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(II) nitrate 2.570 N, cartridge for Digital Titrator

Supplier: Hach
Danger
Synonyms: Mercury dinitrateMercuric nitrate

2393701
97026-486EA 36.69 USD InStock
97026-486
Mercury(II) nitrate 2.570 N, cartridge for Digital Titrator
Mercury (II) nitrate
For determination of Salinity by Digital Titrator titration. 2.570 N. Digits x 0.1 = ppt Salinity in 2 mL sample.

Contains mercury

Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.

Dispose of according to local, state and federal regulations.


Formula: HgN₂O₆
MW: 324.60 g/mol
Melting Pt: -38.9 °C
Density: 4.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011038
CAS Number: 10045-94-0
EINECS: 233-152-3
UN: 2922
ADR: 8 (6.1),II
Merck Index: 12,05933
Order Now
Specification Test Results
Instrument Digital Titrator
Method Name Mercuric Nitrate Titration
Number of tests 40 - 100
Parameter Sulfite
Platform Digital Titrator
Range 0 - 100 ppt Salinity
Titrant 2.570 N Hg(NO3)2

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty