Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.9274 N, cartridge for Digital Titrator

Supplier: Hach
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

1484201 14842-01
97026-238EA 34.3 USD InStock
97026-238 CA14842-01
Sodium hydroxide 0.9274 N, cartridge for Digital Titrator
Sodium hydroxide
Used for volatile acids determination by Digital Titration, 100 - 2400 mg/L Acetic Acid.

  • Accurate results - when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical

Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627
Order Now
Specification Test Results
Method Name Phenolphthalein End Point
Number of tests 40 - 100
Parameter Volatile Acids
Platform Digital Titrator
Range 100 - 2400 mg/L CH3COOH
Titrant 0.9274 N NaOH

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty