Bench Top Sterilizers, Heidolph Tuttnauer
Supplier: HEIDOLPH NA LLC
023210479
89427-576EA 17975.57 USD
89427-576
Bench Top Sterilizers, Heidolph Tuttnauer
Dry Sterilizers Autoclaves (Steam Sterilizers)
Bench Top Sterilizers provide a fast, safe, dependable, and convenient means of sterilizing media, instruments, glassware, clothing, and waste
The line includes a wide range of models from economic manually controlled analog models to electronically controlled models with a built-in printer for GLP-compliant documentation.
Certifications: UL listed. CSA certified.