Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
GreenHouse Plus Parallel Synthesizer, System 28, Heidolph Radleys
Supplier: HEIDOLPH NA LLC
15-85-015-0
80086-226EA 14438.06 USD InStock
80086-226
GreenHouse Plus Parallel Synthesizer, System 28, Heidolph Radleys
Evaporators Parallel Synthesis Systems
The patented GreenHouse Plus Parallel Synthesizer was designed with the help of scientists to perform up to 24 reaction in parallel with a volume range from 0.5 to 7 mL per reaction tube
Its removable reaction block holds 24 glass reaction tubes in the same position as a standard 24-well microplate, enabling the rapid transfer fo samples to and from plates with a multichannel pipette or robotic pipetting system.
Ordering information: This system includes GreenHouse Plus Parallel Synthesizer with head, base, glass cylinder and two reaction blocks, GreenHouse Plus reaction tube, 100pcs, GreenHouse Plus PTFE magnetic stirring bars, 40pcs, GreenHouse Plus and sealing mat evaluation kit, 4cs, Carousel Stirring Hotplate 115V, Carousel temperature controller, Digital thermometer (-250 to 400C) and 200 mm probe, GreenHouse cooling reservoir.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty