Vibramax 100 Platform Shaker, Vibrating, Heidolph North America
Supplier: Heidolph
036130120
82004-942EA 2523.81 USD
82004-942
Vibramax 100 Platform Shaker, Vibrating, Heidolph North America
Shakers and Mixers Orbital Shakers
The Vibramax 100 is a compact and space saving unit with a load capacity of 2 kg
The 3 mm vibration orbit gives excellent results, espcially in large vessels. It allows for smooth to ridged mixing of viscous media and solids. You can set and continuously adjust the variable speed on the analog control knob from 150 to 1,350 rpm. Option attachment with tensions rollers accepts any glassware size. A large selection of attachments is provided for the use of various vessels and sizes. An analog process timer allows for unattended operations and can be set from 1 to 120 min. When the set time has elapsed an acoustic alarm will sound and the operation stops.
Ordering information: Includes Non-skid Rubber Mat.