VWRI89236-532
89236-532
89236-532PK 100.92 USD
89236-532
VWRI89236-532
Nitrate Ionic Strength Adjustor (ISA) for use in nitrate determinations by electrode methods.
  • Non-diluting
  • Accurate determination of nitrate levels
  • Eliminates most interferences
Powder form does not dilute samples. Ideal for determining low level nitrate concentrations. Pack of 100 Powder Pillows.