Fluoride Ionic Strength Adjustor Powder Pillows, VWR®
Supplier: VWR International
89236-530
89236-530PK 102.12 USD InStock
89236-530
Fluoride Ionic Strength Adjustor Powder Pillows, VWR®
Buffers Ionic Strength Adjustment Buffers
For use when making fluoride determinations by electrode methods.
  • Non-diluting
  • Accurate determination of ammonia levels
  • Eliminates most interferences
Eliminates interferences from aluminum and iron. Powder form does not dilute samples. Ideal for determining very low fluoride levels. Use one pillow per 25 mL sample. Pack of 100 Powder Pillows.