Chloride Ionic Strength Adjustor Buffer Powder Pillows, VWR®
89236-528
89236-528PK 88.39 USD
89236-528
Chloride Ionic Strength Adjustor Buffer Powder Pillows, VWR®
For use in chloride determinations by ion-selective electrode methods.
  • Eliminates most interferences
  • Ideal for determining very low chloride levels
Use one pillow per 25 mL sample. Pack of 100 Powder Pillows.