- Home
- Software
- Laboratory Software
- SHUR/Mark® HISTO/Print™ Software, TBS®
SHUR/Mark® HISTO/Print™ Software, TBS®
Supplier: TBS
SM-HP-SL
CA10146-638
SHUR/Mark® HISTO/Print™ Software, TBS®
Software
Laboratory Software
HISTOPrint™ software offers the most comprehensive printing software on the market today and is designed to meet various laboratory needs such as User Screen, Template Design, Print Jobs, Data Filters, Protocols and LIS Output.
It change Field & Barcode Positions and adjust Font Styles and sizes. It flip Text direction, adjust Barcode size, print barcodes in either dot or square formats.
- Network your label, cassette and slide printers to multiple computers
- Enhanced Readability
- LIS Integration
- Change field and barcode positions
- Change font styles
- Adjust font sizes, flip text direction
- Adjust barcode size, Print barcodes in either dot or square
Advanced software is user-definable menu driven software for customized printing of alphanumerical data and load balancing when multiple labelers are integrated. Includes one license for operating one labeler. Included with the purchase of a SHURMark® slide or cassette labeler.
Super software is expansion of advanced software feature-set including 2-D data matrix barcode printing capability and LIS connectivity.
Advanced license to integrate Pre-2013 SHURMark® labelers to TBS HISTOPrint™ software. Each license operates 1 labeling device. Can only be used with advanced software.
Super license operates 1 labeling device. Can only be used with super software.
Once correctly identified, SHUR/Mark® HISTO/Print™ can interpret case information without any additional modifications to existing Laboratory Information Systems (LIS).
Software, Advanced is User-definable menu driven software for customized printing of alphanumerical data and load balancing when multiple labelers are integrated. Includes one license for operating one labeler. Included with the purchase of a TBS SHUR/Mark® Slide or Cassette Labeler.
Software, Super is the expansion of Advanced Software feature-set including 2-D data matrix barcode printing capability and printer load balancing. Accepts extrapolated ASCII formatted patient data from mainframes, networks, & laboratory information systems. Requires some programming by an institution's programmer or LIS vendor. Includes 1 free license capable of operating 1 labeling device. Additional licenses can be purchased. Pre-1992 SHUR/Mark® instruments are not upgradeable and 1992-1995 units may require a hardware upgrade. Advanced Licence is only for use with Advanced Software.
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Inquire for Price
Learn more
About VWR
Item(s) Added to Cart