Diethylene glycol methyl ethyl ether for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
2,5,8-Trioxadecane
, 1-Ethoxy-2-(2-methoxyethoxy)ethane
, Diethylene glycol ethyl methyll ether
M-Clarity™ quality level = MQ200
8.43827.1000
8.43827.0250
8.43827.0250
8.43827.1000
EM8.43827.1000CS
1191
USD
CA8.43827.0250
CA8.43827.1000
EM8.43827.1000
EM8.43827.0250
Diethylene glycol methyl ethyl ether for synthesis, Sigma-Aldrich®
Diethylene glycol methyl ethyl ether
Formula:
C₇H₁₆O₃ MW: 148.2001 g/mol |
MDL Number:
MFCD01727263 CAS Number: 1002-67-1 EINECS: 213-690-5 |
Specification Test Results
Solubility | 76 g/l (25 °C) |
Melting Point | -72 °C |
Molar Mass | 148.2 g/mol |
Flash Point | 82 °C |
Density | 0.925 g/cm3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...